ContactUsLinkComponent
Hi-Vis Yellow Velcro ESD Heel Grounder, Transforming Technologies
Hi-Vis Yellow Velcro ESD Heel Grounder, Transforming Technologies
Catalog # 76243-212
Hi-Vis Yellow Velcro ESD Heel Grounder, Transforming Technologies
Catalog # 76243-212
Supplier Number:  HG1390S
Specifications
  • Color:
    Yellow
  • Cat. No.:
    76243-212
  • Style:
    Cup Style
Specifications
About this item
This heel grounder is constructed from durable materials and features a classic hi-visibility yellow/green hook and loop closure with an elastic insert to maximize fit and comfort and it worn on both feet to provide consistent grounding while in motion.
  • Elastic Hook and Loop closure
  • Buried resistor
  • 1.25″ cup design
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20.