ContactUsLinkComponent
Gauges, Jensen Tools
Gauges, Jensen Tools
Catalog # 76251-394
Supplier:  TESTEQUITY LLC TE
Gauges, Jensen Tools
Catalog # 76251-394
Supplier:  TESTEQUITY LLC TE
Supplier Number:  ATG36Z-S
Specifications
  • Description:
    Rorque Gauge
  • Cat. No.:
    76251-394
  • Torque:
    0.6 - 2.8" oz.
Specifications
About this item
Analog handheld torque gauge ide for measuring and tightening in low torque ranges.
  • Three Fingered Keyless Metal Chuck
  • CW and CCW measuring with side and top memory pointers
Delivery: Calibration Certificate Included.