Wrist Strap Monitor, SCS, Desco Industries
Catalog # 76240-014
Supplier: Desco
Specifications
- Description:Wrist Strap Monitor, Portable
- Cat. No.:76240-014
Specifications
About this item
This wrist strap monitor visual and audible alarms indicate proper grounding of the operator and ground point.
Monitor can provide ground to a portable worksurface when connected through the 10 mm snap.
Certifications: Calibrated with NIST.