Reversible/Stacking Ground Cord, Menda
76242-580
: Desco
- Description:10' Ground Cord, 1 Meg Resistor
- Cat. No.:76242-580
Reversible/stacking ground cord fits either 10 mm stud or socket on mat.
- Stacking snap allows it to be used as a common point ground