Order Entry
United States
Orders LinkContactUsLinkComponent
Precision Nut Driver, Wiha Tools

Precision Nut Driver, Wiha Tools

Precision Nut Driver, 1/4", 6.7"
Catalog # 76242-314
Supplier:   WIHA TOOLS TE
Supplier #:  26563
Precision Nut Driver, Wiha Tools
Catalog # 76242-314
Supplier:  WIHA TOOLS TE
Precision Nut Driver, Wiha Tools

Precision Nut Driver, 1/4", 6.7"

Price
$33.03
Log in for availability
Each
Product Details & Documents

This exact fit narrow profile precision machined nut driver is made from CRM-72 tool steel.

  • Free turning finger control cap
  • Blade length: 2.4"

Hard chromed finished for best corrosion protection.

Certifications: RoHS compliant.

Specifications
  • Description:
    Precision Nut Driver, 1/4"
  • Overall length:
    6.7"