Order Entry
United States
ContactUsLinkComponent
LED Light, Production Basics
LED Light, Production Basics
Catalog # 76253-280
Supplier:  Production Basics
undefined
LED Light, Production Basics
Catalog # 76253-280
Supplier:  Production Basics
Supplier Number:  PB-8900

Specifications

  • Description:
    LED Overhead Light, Includes 2×24" Bulbs
  • Cat. No.:
    76253-280
  • Diameter:
    48"

Specifications

About this item