ContactUsLinkComponent
Aerostat® Ionizing Blower, Simco Ion
Aerostat® Ionizing Blower, Simco Ion
Catalog # 76246-960
Supplier:  Simco
Aerostat® Ionizing Blower, Simco Ion
Catalog # 76246-960
Supplier:  Simco
Supplier Number:  4011009
Specifications
  • Description:
    FPD wide coverage ionizing blower
  • Cat. No.:
    76246-960
  • W×D×H:
    1215x160x100 mm (47.75x6.25x4")
  • Electrical:
    120 V, 60 Hz
Specifications
About this item
Aerostat FPD four fan ionizing blower is designed to control electrostatic charges in assembly and electronics manufacturing.
  • Extremely fast static discharge
  • Built-in emitter point cleaner for simplified maintenance
  • Ion balance and ion output LED provide real-time ionization status
  • Emitter Points never need replacement
The Aerostat FPD Ionizing Air Blower provides superior fast neutralization over an extended surface area. Available in two, three and four-fan models, this high velocity overhead blower provides superior coverage up to 5'.