ContactUsLinkComponent
Antistatic Utility Tape Dispenser 620, with base, 3M™
Antistatic Utility Tape Dispenser 620, with base, 3M™
Catalog # 76247-892
Supplier:  3M
Antistatic Utility Tape Dispenser 620, with base, 3M™
Catalog # 76247-892
Supplier:  3M
Supplier Number:  7100049758
Specifications
  • Description:
    Antistatic utility tape dispenser 620, with base
  • Cat. No.:
    76247-892
Specifications
About this item
Tape dispenser 620 is designed for use in ESD protected areas. The dispenser is made of conductive and static-dissipative materials which will minimize or eliminate problems associated with conventional dispensers.
  • Minimizes problems with static-dissipative and conductive material construction
  • Minimizes floor space requirements
Stainless-steel blade easily cuts the tape with maximum strength.