Mycoplasma Agar
Catalog # 76300-102
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Environmentally Preferable:
- Cat. No.:76300-102
- Plate size:15×60 mm
Specifications
About this item
For the cultivation of Mycoplasma and the detection of Mycoplasma contamination in biologics and biological processes.
- Optically clear polystyrene - For distortion free viewing.
- Heavy-Duty - Reduces breakage during shipping.
- Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
Ordering information: Minimum order: 5 Packs
Packaging: Breathable Packaging - Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture.