Indent Crimp Tool, DMC
Catalog # 76253-236
Supplier: DMC TE
Specifications
- Description:Crimp Tool
- Cat. No.:76253-236
Specifications
About this item
This crimp tool is a fast, manually operated tool for crimping AS7928 (MIL-T-7928) ring terminals and splices for wire sizes 26 through 14.
- Cycle controlled
- Adjustable insulation barrel
- Equipped with a locator to hold terminals and splices in place for crimping
This crimp tool provides a ratchet-controlled crimp to ensure full crimp force/closure is applied. The handles will not open until they have been fully closed and the crimp completed.