Order Entry
United States
ContactUsLinkComponent
Plastic ESD Boxes, Conductive Containers
Plastic ESD Boxes, Conductive Containers
Catalog # 76253-746
Plastic ESD Boxes, Conductive Containers
Catalog # 76253-746
Supplier Number:  24R-1039

Specifications

  • Description:
    Black
  • Dimensions:
    1/2×29/16×1"
  • Cat. No.:
    76253-746
  • Weight:
    6 lbs.

Specifications

About this item

Injection molded conductive plastic boxes feature a new design that provides extra durability with dependable ESD protection.

  • Extra strong hinges and latches
  • Tongue and groove closure
  • Thicker, stronger walls
Packaging: Pack of 50/Case.