Amp-TE Connectivity Ratchet Crimp Tool, Newark/Element
Catalog # 76251-548
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Ratchet Crimp Tool
- Environmentally Preferable:
- Cat. No.:76251-548
Specifications
About this item
This T-head hand crimp tool is designed for AMP PIDG/STRATO-THERM/PLASTI 26-14AWG terminals and splices.
- The handle of the crimp tool is color-coded to match the color coding of the product to be applied
- PLASTI-GRIP terminals on solid or stranded copper wire sizes 22 through 14AWG
- STRATO-THERM terminals on stranded copper wire sizes 26 through 14AWG
The insulation adjustment indicator is used to control crimp height of the insulation barrel. It can be set at any of four positions corresponding to insulation diameter.
The CERTI-CRIMP hand crimping tool ratchet control ensures full crimping of the product. Once engaged, the ratchet will not release until the handles have been fully closed.