

Hydrochloric acid 0.01M, sterile-filtered
Catalog # 103701-214
Supplier: ADVANCED BIOMATRIX, INC. MS
CAS Number:
Specifications
- Size:100 ml
- Cat. No.:103701-214
Specifications
About this item
:
:36.46
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324