Order Entry
United States
ContactUsLinkComponent
 
Fluorescent Blub, Florida Bulb & Ballast
  76246-576
Fluorescent Blub, Florida Bulb & Ballast
  76246-576
 :  F30T8/BLB

 

  • Description:
    Black Light Fluorescent Bulb, 36"
  • Cat. No.:
    76246-576
  • Power:
    30 W

 

 

Fluorescent blub 30 W T8 black 36".