Tape 8901, Polyester, 3M
Supplier: 3M
Certificates
About this item
Polyester Tape 8901 is a thin, conformable polyester tape ideal for masking in high temperature applications and adheres well to silicone coated surfaces.
- Thermal and chemical resistance while maintaining flexibility
- Clean removal during high-temperature masking operations
The translucent blue backing is tear resistant. Coated with a silicone adhesive, this tape functions well in masking for powder coating to provide sharp paint lines. It removes cleanly and in one-piece from most surfaces, avoiding slivering and breaking. It also excels in permanent splicing operations on silicone liners, papers and films subject to high temperatures.
Frequently Bought Together