GripTip Pipette Tips, Racked, Integra Biosciences
Supplier: INTEGRA MS
Certificates
About this item
The GripTips are comfortably loaded and ejected without any physical effort due to a power assisted mechanism activated by a push button. The loading procedure also guarantees that GripTips are not touched by the user and remain sterile.
- Extended length design enabling easy access in to labware types such as tubes and deep well blocks
- Available in sterile and sterile filtered configurations to serve all application requirements
The tip racks are reinforced to guarantee proper loading of the GripTips.
Filtered GripTips reliably protect the pipetting head and samples against contaminations caused by aerosols.
Frequently Bought Together