Polyester Overlaminate Labels, WorkHorse®, Brady
General purpose overlaminate that withstands high temperatures, dirt, oil and cleaning solvents while displaying excellent clarity.
- Ideal for use on PC boards
- Chemical and corrosion resistant
- Humidity, UV and weather resistant
The Die-Cut overlaminate is made of clear polyester material.
General purpose overlaminate that withstands high temperatures, dirt, oil and cleaning solvents while displaying excellent clarity.
- Ideal for use on PC boards
- Chemical and corrosion resistant
- Humidity, UV and weather resistant
The Die-Cut overlaminate is made of clear polyester material.
Product Family Options
- DescriptionMaterialColorDimensions
- Overlaminate labels, glossy finishPolyesterClear5.1×16.5 mm (0.2×0.65")-: -
- Die-cut overlaminate labels, glossy finishPolyesterClear6.4×25.4 mm (0.25×1")Each$667.70: 16101
- Overlaminate labels, glossy finishPolyesterClear12.7×25.4 mm (0.5×1")-: -