ContactUsLinkComponent
Magnum FLX® Enhanced Universal Magnet Plate, Alpaqua Engineering LLC MS
Magnum FLX® Enhanced Universal Magnet Plate, Alpaqua Engineering LLC MS
Catalog # MSPP-A000400
Magnum FLX® Enhanced Universal Magnet Plate, Alpaqua Engineering LLC MS
Catalog # MSPP-A000400
Supplier Number:  A000400
Specifications
  • Description:
    Enhanced Universal Magnet Plate
  • Cat. No.:
    MSPP-A000400
Specifications
About this item
The Magnum FLX® enhanced universal magnet plate contains proprietary solid-core ring magnets for increased speed and performance.
  • Robotic platforms
  • Unique geometry of the solid-core magnets
  • Rapid separation of samples up to 2 ml in volume and elution in as little as 10 µl
Universal compatibility with all commonly used 96-well microplates.