Membrane Filters, Polycarbonate, Sterlitech
Catalog # MSPP-PCT301310
Supplier: STERLITECH MS
Specifications
- Ø:13 mm
- Pore size:3.00 µm
- Environmentally Preferable:
- Cat. No.:MSPP-PCT301310
- Supplier no.:PCT3013100
Specifications
About this item
Polycarbonate Track Etch (PCTE) membranes are made from a thin, microporous polycarbonate film material.
Ideal for use in blood assays and high-purity and general filtration.