Orders LinkContactUsLinkComponent
Scissors, Sharp, Fine Science Tool
Scissors, Sharp, Fine Science Tool
Marketsource
Scissors, Sharp, Fine Science Tool
Marketsource
Certificates
Scissors, Sharp, Fine Science Tool
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Scissors made of stainless steel material.
  • Sharp tips
  • Non-serrated
Product Family Options
Product Information
  • Length
    Material
    Blade length
    Blade type
    Curvature
    Availability
    Price
  • 9.0 cm
    Stainless steel
    22 mm
    Sharp/sharp, Non-serrated
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $145.38
    $145.38
    Each
       $145.38
    Supplier Number: 14060-09
    Specifications:
    Marketsource
    Marketsource
  • 10.5 cm
    Stainless steel
    21 mm
    Sharp/sharp, Non-serrated
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $145.38
    $145.38
    Each
       $145.38
    Supplier Number: 14060-10
    Specifications:
    Marketsource
    Marketsource