Order Entry
United States
ContactUsLinkComponent
Vinyl Caps, VC and VCF Series, Caplugs
Vinyl Caps, VC and VCF Series, Caplugs
Catalog # 76243-716
Supplier:  Caplugs
Vinyl Caps, VC and VCF Series, Caplugs
Catalog # 76243-716
Supplier:  Caplugs
Supplier Number:  VCF-1000-24

Specifications

  • Description:
    Vinyl Cap with Flanges
  • Height:
    1.5"
  • Environmentally Preferable:
  • Cat. No.:
    76243-716
  • Cap color:
    Black
  • Nominal diameter:
    1"

Specifications

About this item

VCF Series caps are designed for easy removal with a flange at the closed end large enough to grasp, yet small enough to not interfere with the application.

  • Wide range of sizes
  • Easy to use
  • Vast color range available