Order Entry
United States
ContactUsLinkComponent
Auto-Feed Soldering station, JBC Tools USA, JBC
Auto-Feed Soldering station, JBC Tools USA, JBC
Catalog # 76245-754
Supplier:  Avantor
Auto-Feed Soldering station, JBC Tools USA, JBC
Catalog # 76245-754
Supplier:  Avantor

Specifications

  • Temperature range:
    90...450 °C (190...840 °F)
  • Description:
    Auto-Feed soldering station, 5.81 kg
  • Dimensions:
    195×200×240 mm
  • Environmentally Preferable:
  • Cat. No.:
    76245-754

Specifications

About this item

Auto-Feed soldering station is the ideal solution for extensive soldering applications or when a free hand is required. The AL station automates the soldering process by feeding solder wire.

  • Possible to select continuous or discontinuous operation modes

AL station works with C250 cartridges.