ContactUsLinkComponent
Mini Coax Cable Stripper, UST-185, Jensen Tools + Supply
Mini Coax Cable Stripper, UST-185, Jensen Tools + Supply
Catalog # 76252-256
Supplier:  TESTEQUITY LLC TE
Mini Coax Cable Stripper, UST-185, Jensen Tools + Supply
Catalog # 76252-256
Supplier:  TESTEQUITY LLC TE
Supplier Number:  JON-UST-185
Specifications
  • Description:
    Mini Coax Cable Stripper
  • Cat. No.:
    76252-256
Specifications
About this item
The UST-185 mini coax cable stripper is specifically designed to provide a 1/4×1/4" two step strip on dual and quad shield mini coax cables.
  • Capable of over 5000 strips
This tool works on Belden, Commscope, Times Fiber and other mini coax cables. The unique blade design has blades setup for Dual shield on one side and Quad shield on the other.