Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bags
Storage Bags
Fluorescent Storage Bags, Associated Bag
Fluorescent Storage Bags, Associated Bag
Fluorescent Storage Bags, Associated Bag
Catalog # 76249-566
Supplier:
Associated Bag
Expand image
Fluorescent Storage Bags, Associated Bag
Catalog # 76249-566
Supplier:
Associated Bag
Supplier Number:
SP-170-0151095
Specifications
Color:
Orange
Description:
Fluorescent lable scarp, 2×3"
Cat. No.:
76249-566
Specifications
About this item
Product Details
Product Details