Anti-Static Cleanroom Marking Tapes, UltraTape Industries
Supplier: Ultratape Industries
Certificates
About this item
Cleanroom Anti-static tape available in for floor, tool and safety marking as well as a wide range of applications were opaque colors are needed. Printed and overlaminated for extra durability and solvent resistance. Bright opaque colors and black printed text.
- Excellent durability
- Anti-static and cleanroom compatible
- Solvent resistant
Frequently Bought Together