Order Entry
United States
ContactUsLinkComponent
Worksurfaces, Production Basics
Worksurfaces, Production Basics
Catalog # 76253-428
Supplier:  Production Basics
Worksurfaces, Production Basics
Catalog # 76253-428
Supplier:  Production Basics
Supplier Number:  PB-7936

Specifications

  • Worksurface material:
    16 Gauge Stainless Steel
  • Description:
    Worksurfaces, Wrap Edges
  • Cat. No.:
    76253-428
  • W×D:
    48×30"
  • Weighing capacity:
    66 lbs.

Specifications

About this item

Durable and clean stainless steel worksurfaces are ideal for packaging and cleanroom applications.