Adhesive Plate Sealers, EdgeBio
Catalog # MSPP-48461_U
Supplier: Edge Biosystems
Specifications
- Description:Adhesive plate sealers
- Cat. No.:MSPP-48461_U
- Supplier no.:48461
Specifications
About this item
These adhesive plate sealers can be used for protecting samples in 96- well and 384- well plates during shaking, incubation, and storage.
- Resistant to heat and cold (−20 to 100 °C)
These adhesive plate sealers can be secured to any flat surface to eliminate waste and clutter.
Packaging: Packed in sets of 25 sealers.