Orders LinkContactUsLinkComponent
Filter Flasks, Advantec MFS
Marketsource
Supplier:   Advantec MFS
Filter Flasks, Advantec MFS
Supplier:  Advantec MFS
Marketsource
Certificates
Filter Flasks, Advantec MFS
4 Options Available Below
Product Details & Documents
These borosilicate filter flasks and silicone rubber stoppers are compatible with Advantec filter holders.
  • Side arm connection 3/8"
  • Flasks with ground glass joint fit holders with female joint (GFA, P/FC 2)
These flasks can be used to collect filtrate or to act as a trap to prevent liquid from damaging pump or vacuum line.
#5 and #8b stoppers adapt 13 mm, 25 mm and KP 47 filter holders to 125 ml (#5) and 1 l (#8b) flasks. #8 stopper adapts 47 mm (except KP 47) and 90 mm filter holders to the 1 l flask.
Product Family Options
Product Information
  • Capacity
    [ST] joint no.
    Availability
    Price
  • 101104-592
    Filter Flask
    Filter Flask
    1 L
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $285.72
    $285.72
    Pack of 1
       $285.72
    Supplier Number: 311430
    Specifications:
    Marketsource
    Marketsource
  • 101104-936
    Filter Flask
    Filter Flask
    250 ml
    19/22
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $285.03
    $285.03
    Pack of 1
       $285.03
    Supplier Number: 311225
    Specifications:
    Marketsource
    Marketsource
  • 101104-174
    Filter Flask
    Filter Flask
    1 L
    40/35
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $319.06
    $319.06
    Pack of 1
       $319.06
    Supplier Number: 311425
    Specifications:
    Marketsource
    Marketsource
  • 101104-128
    Filter Flask
    Filter Flask
    2 L
    40/35
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $463.58
    $463.58
    Pack of 1
       $463.58
    Supplier Number: 311426
    Specifications:
    Marketsource
    Marketsource
Stoppers
Product Information
  • Catalog #
    Description
    Details
    Fits
    Availability
    Price
  • Silicone rubber stopper, #8b
    -
    KP 47
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $27.17
    $27.17
    Pack of 1
       $27.17
Product Information
  • Catalog #
    Description
    Details
    Fits
    Availability
    Price
  • Silicone rubber stopper, #8
    -
    P/FC-1
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $26.63
    $26.63
    Pack of 1
       $26.63