Go/No Go Plug Gage Assembly, Zoro
Catalog # 76250-088
Supplier: ZORO - CUSTOMER SERVICE TE
Specifications
- Description:Plug for use with thread size: #4 - 40,
- Length:2²¹⁄₃₂"
- Cat. No.:76250-088
Specifications
About this item
Go/No Go, Taperlock thread plug gage, Class 2B.
- Made of tool steel
- Chrome plated finish
- Tolerance Class X
- UNC Thread
For calibration of measuring equipment or to gage parts directly.
1.75" (handle)
Delivery: Includes Certificate of Accuracy Traceable to NIST.