Specifications
- Antibody Type:Primary
- Antigen Name:Transferrin Fluorescein Conjugated
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:25 µL
- Shipping Temperature:Dry Ice
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- Cat. No.:75933-052
- Supplier no.:209-4234S
Specifications
About this item
Human transferrin is encoded by the TF gene and is an iron-binding blood plasma glycoprotein that controls the level of free iron in biological fluids. Human transferrin binds iron very tightly but reversibly. Human transferrin is the most important iron pool in mammals. Human transferrin has a molecular weight of around 80 kDa and contains 2 specific high-affinity Fe(III) binding sites. The affinity of Human transferrin for Fe(III) is extremely high but decreases progressively with decreasing pH below neutrality. Human Transferrin also plays a role in the immune system, creating environments low in iron for which many pathogenic bacteria are unable to thrive.
Anti-Human transferrin fluorescein is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Type: Primary
Antigen: Transferrin Fluorescein Conjugated
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity: