Specifications
- Antibody Type:Primary
- Antigen Symbol:WFDC2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75933-632
- Supplier no.:600-401-FY9
Specifications
About this item
The WAP four-disulfide core domain protein 2 (WFDC2), also known as epididymal protein 4, is a member of the WFDC domain family, a family of proteins that is characterized by the presence of Whey Acidic Protein (WAP) domain, and is highly expressed in the lung and salivary gland (1,2). Members of this family include SLPI and elafin, antiproteinases involved in the innate immune system (2). WFDC2 has been proposed to play a critical role in tumor formation and growth in ovarian cancer cells through the regulation of growth- and apoptosis-associated genes and may thus be a potential therapeutic target for epithelial ovarian cancer (3).
Anti-WFDC2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 13 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: WFDC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: