Specifications
- Antibody Type:Primary
- Antigen Symbol:Wdr5
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µL
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:multi-step process which includes delipidation and salt fractionation
- Cat. No.:75933-614
- Supplier no.:100-401-V89
Specifications
About this item
Wdr5 belongs to the family of WD repeat proteins which are involved in different cellular processes such as cell cycle progression, signal transduction, apoptosis, and gene regulation. It is a component of the Set1A and Set1B histone H3 methylation complexes. These complexes methylate lysine 4 of H3, thereby activating gene transcription. Wdr5 interacts with H3 dimethyl K4, but not with tri or mono methylated H3K4. Anti-Wdr5 Antibody is ideal for research in Cell Cycle Control, Signal Transduction, Apoptosis, Epigenetics, Gene Expression and Cancer.
Anti-Wdr5 Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 36 kDa in the appropriate cell lysate or extract.
Type: Primary
Antigen: Wdr5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: