Order Entry
United States
ContactUsLinkComponent
 
Anti-STRADB Rabbit Polyclonal Antibody
Catalog # 75928-932
undefined
Anti-STRADB Rabbit Polyclonal Antibody
Catalog # 75928-932
Supplier Number:  600-401-BY1

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    STRADB
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Storage Temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping Temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. No.:
    75928-932
  • Supplier no.:
    600-401-BY1

Specifications

About this item

ILPIP (ILP-interacting protein) has recently been shown to interact with XIAP, a member of the IAP (Inhibitor of Apoptosis) protein family. Together, these two proteins synergistically activate the TAK1/JNK1 signal transduction pathway, which acts to protect against the interleukin-1b converting enzyme of Fas-induced apotosis. This protein was independently isolated and characterized as polyploidy-associated protein kinase (PAPK), a member of the Ste20/germinal center kinase family that modulates cytoskeletal organization and cell survival, suggesting that its protective role may be a function of its kinase activity. Although two mRNA sequences have been isolated, only the shorter isoform has been observed.

Anti-ILPIP Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 47 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: STRADB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: