- Antibody Type:Primary
- Antigen Name:QKI-5
- Antigen Symbol:QKI-5
- Clonality:Monoclonal
- Clone:S147-6
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75931-424
- Supplier no.:200-308-W79
QKI is also called Protein Quaking or HqkI. QKI is an RNA-binding protein that plays a central role in myelinization. QKI acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation, and is itself, regulated by alternative splicing. QKI is expressed in the frontal cortex of brain, but is shown to be downregulated in the brain of schizophrenic patients. Anti-QKI (pan) is ideal for research in Neuroscience and Gene Expression.
Anti-QKI (pan) R-PE Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~36-38kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: QKI-5
Clonality: Monoclonal
Clone: S147-6
Conjugation: RPE
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: