Specifications
- Antibody Type:Primary
- Antigen Symbol:MATN4
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75929-868
- Supplier no.:600-401-CR6
Specifications
About this item
Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. Members of the matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices such as embryonic kidney, lung, bone, cartilage and nervous system. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation. MATN4 could serve as an odontoblast differentiation marker, e.g. in odontoblast stem cell research.
Anti-MATN4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 68 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: MATN4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: