Specifications
- Antibody Type:Primary
- Antigen Symbol:IL37
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75928-924
- Supplier no.:600-401-BX7
Specifications
About this item
Interleukin 37 (IL-37) is a member of the interleukin 1 cytokine family (reviewed in 1). It can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). IL-37 also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18 (1). IL-37 can also interact with Smad3 and expression of IL-37 in macrophages or epithelial cells resulted in near complete suppression of pro-inflammatory cytokines, suggesting that IL-37 may act as a natural suppressor of the innate inflammatory and immune responses (2,3).
Anti-IL-37 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 24 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: IL37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: