- Antibody Type:Primary
- Antigen Symbol:DsRed
- Clonality:Monoclonal
- Clone:8E5.G7
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Western Blot:Yes
- Size:25 µL
- Shipping Temperature:Dry Ice
- Purification:Protein A purified
- Cat. No.:75931-550
- Supplier no.:200-301-379S
Antibodies to RFP (Discosoma spp.) are intended for use in immunological assays including ELISA, western blotting, fluorometry and fluorescence activated cell sorting (FACS). RFP Proteins are useful markers for imagining protein localization,
monitoring physiological processes, and detecting transgenic expression. Rockland's anti-RFP antibody can be used to detect native RFP and RFP variants.
Anti-RFP antibodies is designed to detect Red Fluorescent Protein and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen. Biotin conjugated anti-RFP used in a sandwich ELISA with unconjugated anti-RFP is well suited to titrate RFP in solution. The detection antibody conjugated to biotin is subsequently reacted with streptavidin conjugated HRP (code # S000-03). Fluorochrome conjugated anti-RFP can be used to detect RFP by immunofluorescence microscopy in cell expression systems and can detect RFP containing inserts. Significant amplification of signal is achieved using fluorochrome conjugated anti-RFP relative to the fluorescence of RFP alone. For immunoblotting use either alkaline phosphatase or peroxidase conjugated anti-RFP to detect RFP or RFP containing proteins on western blots. Optimal titers for applications should be determined by the researcher.
Type: Primary
Antigen: DsRed
Clonality: Monoclonal
Clone: 8E5.G7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: