Specifications
- Antibody Type:Primary
- Antigen Symbol:Apba1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75933-454
- Supplier no.:600-401-FV4
Specifications
About this item
Upregulated obese product 11 (UROP11) was isolated from subtractive library for mouse hypothalamus. It is detected in numerous tissues and is upregulated in the hypothalamus and adipocytes in obese mice. UROP11 is also observed in human neural and lymphoid tissues; its expression appears to be regulated by the level of lymphoid activation. Urop11 expression is strongly upregulated both in vivo in mouse hypothalamus and in vitro in mouse neural cell lines after treatment with leptin, indicating Urop11 is a target of leptin activity and thus may play a role in obesity.
Anti-UROP11 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 93 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: Apba1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: