$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Bags
Storage Bags
Static Shield Bag, Z-Mar Technology
Static Shield Bag, Z-Mar Technology
76244-768
:
Z-MAR TECHNOLOGY INC. TE
Static Shield Bag, Z-Mar Technology
76244-768
:
Z-MAR TECHNOLOGY INC. TE
:
025-0184-040
Description:
Static shielding bag, cushioning, ESDS
Dimensions:
10×12"
Cat. No.:
76244-768