Chicago Latrobe® Jobber Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
Certificates
About this item
These general-purpose jobber length drills are designed for drilling a wide variety of materials on different range of operating conditions.
- Made of premium high-speed steel
- 118°-point style
- Extra rigid for portable drilling
- Available in fractional, letter and wire gage size
Black oxide surface finish increases wear resistance and adds lubricity, improving chip flow.
Frequently Bought Together