L(+)-Aspartic acid 98.5-101.5% (dried basis) USP, J.T.Baker®
Supplier: Avantor
MOC: Regulated
About this item
CAS Number:56-84-8
:
Formula:C₄H₇NO₄
MDL Number:MFCD00002616
:133.1 g/mol
:Management of Change (MOC) category = R
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.02 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤10 ppm
- USP - Related Compounds: Maleic Acid:≤0.05 %
- USP - Related Compounds: Malic Acid:≤0.20 %
- USP - Related Compounds: Fumaric Acid:≤0.10 %
- USP - Related Compounds: Any unspecified impurity:≤0.05 %
- USP - Related Compounds: Total unspecified impurities:≤0.10 %
- USP - Specific Rotation [Å]²º^D:24.0 - 26.0 Degree
- USP - Loss on Drying:≤0.5 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
spec_JT6254-7
Product specification
Frequently Bought Together