ContactUsLinkComponent
Tool Extractor, Jensen Tools
Tool Extractor, Jensen Tools
Catalog # 76245-298
Supplier:  TESTEQUITY LLC TE
Tool Extractor, Jensen Tools
Catalog # 76245-298
Supplier:  TESTEQUITY LLC TE
Supplier Number:  EX5
Specifications
  • Description:
    PLCC Extractor
  • Cat. No.:
    76245-298
Specifications
About this item
These extracter use to rmeove the chip, range of PLCC sizes from 20 pins through 84 pins from any socket without damage to the chip.
  • ESD safe handles for use on all static safe components
  • Made of stainless steel
Spring loaded one-hand design requires no pulling, just squeeze handles and chip is lifted from the socket.