Dispensing Tips, OK International
About this item
The tapered tip series designed with a double helix threads and universal luer lock to provide secure connection to industrial syringes or dispense valves.
- Silicone and chloride-free with a maximum operating pressure 100 psi (6.9 bar)
- Flash free orifice ensures accurate and repeatable dispensing
These tips are recommended for applications where dispensing medium and high-viscosity fluids with a high degree of precision and consistency is required. A smooth internal fluid path designed reduces turbulence flow, prevents air entrapment and fluid dripping caused by back pressure built up.
7 Options Available Below
Product Details & Documents
The tapered tip series designed with a double helix threads and universal luer lock to provide secure connection to industrial syringes or dispense valves.
- Silicone and chloride-free with a maximum operating pressure 100 psi (6.9 bar)
- Flash free orifice ensures accurate and repeatable dispensing
These tips are recommended for applications where dispensing medium and high-viscosity fluids with a high degree of precision and consistency is required. A smooth internal fluid path designed reduces turbulence flow, prevents air entrapment and fluid dripping caused by back pressure built up.
Product Family Options
Product Information
- GaugeLengthColorAvailabilityPrice
- 76239-416Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor1431.8 mm (1.25")PinkLog in for availabilityPack of 50$100.45Supplier Number: TT14-DHUV-PK
- 76239-556Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor1631.8 mm (1.25")GreyLog in for availabilityPack of 50$120.00Supplier Number: TT16-DHUV-PK
- 76239-418Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor1831.8 mm (1.25")GreenLog in for availabilityPack of 50$94.16Supplier Number: TT18-DHUV-PK
- 76239-420Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor2031.8 mm (1.25")PinkLog in for availabilityPack of 50$93.05Supplier Number: TT20-DHUV-PK
- 76239-422Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor2231.8 mm (1.25")BlueLog in for availabilityPack of 50$98.45Supplier Number: TT22-DHUV-PK
- 76239-424Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor2531.8 mm (1.25")RedLog in for availabilityPack of 50$78.49Supplier Number: TT25-DHUV-PK
- 76240-408Dispensing poly tip, tapered, with UV inhibitorDispensing poly tip, tapered, with UV inhibitor2731.8 mm (1.25")ClearLog in for availabilityPack of 50$115.53Supplier Number: TT27-DHUV-PK