Order Entry
United States
ContactUsLinkComponent
 
Tris HCl pH 9.0, 1.0 M
Catalog # 76236-404
CAS Number:  
Tris HCl pH 9.0, 1.0 M
Catalog # 76236-404
Supplier Number:  MB-028-1000
CAS Number:  

Specifications

  • Description:
    1.0 M Tris HCl pH 9.0
  • Size:
    1 L
  • Cat. No.:
    76236-404

Specifications

About this item

Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology.

  • This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.
  • This buffer consists of 1.0 M Tris HCl at pH 9.0.

In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.

Application Note: Tris solution is suitable for molecular immunology, molecular biology and nucleic acid methodologies, when applicable. Visit our newly expanded web site at www.rockland-inc.com for methods using this and other buffers. This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 1.0 M Tris HCl at pH 9.0.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade (WFI) water.