Specifications
- Description:Blocking Buffer For Immunohistochemistry Serum And Azide Free
- Size:100 mL
- Cat. No.:RLMB-071-0100
- Supplier no.:MB-071-0100
Specifications
About this item
Protein blocker specifically designed for immunohistochemical staining where detection occurs using an immunoenzymatic reaction with substrate, i.e. peroxidase (HRP) with diaminobenzidine (DAB)
- Specifically inhibits non-specific staining of IHC detection reagents
- Undiluted or 1:1 in TBS
- The buffer is pH adjusted to pH 7.6.
A detailed protocol for immunohistochemical staining is available at www.rockland-inc.com.
Application Note: This immunohistochemistry protein blocker is specifically designed for immunohistochemical staining where detection occurs using an immunoenzymatic reaction with substrate, i.e. peroxidase (HEP) with diaminobenzidine (DAB). This product specifically inhibits non-specific staining of IHC detection reagents. It may be used on various specimens, including cultured cells and primary tissue to lower background staining. Optimum performance is achieved using this product undiluted. However, Immunohistochemistry Protein Blocker can often be diluted 1:1 in TBS without a significant loss of performance. This immunohistochemistry protein blocker consists of proprietary blocking agents and stabilizers prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI).