Compartment Box, Flambeau Inc.
Catalog # 76248-250
Supplier: FLAMBEAU INC. TE
Specifications
- Color:Black
- Dimensions:123/4× 81/2×21/8" (32.39×21.59×5.40 cm) (Compartments)
- Cat. No.:76248-250
Specifications
About this item
The 600/800 series conductive box reduces electrical resistance and dissipates static-charge buildup to alleviate electrostatic discharge (ESD).