Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
:120-47-8
:
:C₉H₁₀O₃
:MFCD00002353
:Management of Change (MOC) category = R
:166.18 g/mol
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- NF - Assay (C₉H₁₀O₃):98.0 - 102.0 %
- NF - Appearance (white, crystalline powder):Passes Test
- NF - Color of Solution:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Acidity:Passes Test
- NF - Related Substances - p-Hydroxybenzoic Acid:≤0.5 %
- NF - Related Substances - Unspecified Impurities (each):≤0.5 %
- NF - Related Substances - Total Impurities:≤1.0 %
- NF - Residue on Ignition:≤0.1 %
- EP/BP - Assay (C₉H₁₀O₃):98.0 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Related Substances - Impurity A:≤0.5 %
- EP/BP - Related Substances - Unspecified Impurities (each):≤0.5 %
- EP/BP - Related Substances - Total Impurities:≤1.0 %
- EP/BP - Sulfated Ash:≤0.1 %
- JP - Acidity:Passes Test
- JP - Assay:98.0 - 102.0 %
- JP - Clarity and Color of Solution:Passes Test
- JP - Identification:Passes Test
- JP - Melting Point:115 - 118 °C
- JP - Heavy Metals:≤20 ppm
- JP-RelatedSubstances- p-Hydroxybenzoic Acid:≤0.5 %
- JP-Related Substances-Unspecified Impurities (each):≤0.5 %
- JP-Related Substances-Total Impurities:≤1.0 %
- JP - Residue on Ignition:≤0.10 %
Frequently Bought Together