To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:PE (Phycoerythrin)
- Source:
- Species:Human
- Size:50 tests
- Environmentally Preferable:
- Protein/Peptide Name:HLA-A*23:01
- Cat. No.:103500-644
MHC Tetramers are used for the detection of antigen-specific T-cell populations.
- Class-I
MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.