QuickSwitch™ H-2 Kb Tetramer Kit-BV421, MBL International
Catalog # 103500-882
Supplier: Avantor
CAS Number:
Specifications
- Size:25 μg
- Species Reactivity:Mouse
- Cat. No.:103500-882
Specifications
About this item
QuickSwitch™ Tetramer Kit utilizes a patented technique for exchanging up to ten peptides on an MHC class I tetramer. Reduce time and cost with this kit to create your own custom tetramers. The resulting custom tetramers can be used for multiple applications, including screening immunogenic peptides.
Research Area: Immunology, Immune Monitoring