Specifications
- Description:HUMAN BLOOD in Alsevers -
- Size:50 mL
- Shipping Temperature:Wet Ice
- Cat. No.:RLR314-0050
- Supplier no.:R314-0050
Specifications
About this item
HUMAN BLOOD in Alsevers -
- aseptically collected and prepared to order
- blood draw in Anticoagulant
Human whole blood is aseptically collected and processed upon receipt of your order. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Human red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.